Learn More
Aviva Systems Biology Corp PSMD4 Antibody - C-terminal region (ARP32341_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32341_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSMD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PSMD4. Alternate names: AF, ASF, S5A, AF-1, MCB1, Rpn10, pUB-R5ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-54534