Learn More
Aviva Systems Biology Corp PSMD8 Antibody - C-terminal region (ARP48216_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48216_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSMD8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PSMD8. Alternate names: HIP6, HYPF, MGC1660, Nin1p, Rpn12, S14, p31ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59762