Learn More
Aviva Systems Biology Corp PSMD9 Antibody - C-terminal region (ARP59344_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59344_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSMD9. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human PSMD9. Alternate names: MGC8644, Rpn4, p27ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-668