Learn More
Sino Biological PSME2 / PA28b Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 14640MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PSME2 / PA28b (rh PSME2 / PA28b; Catalog#14640-H07E; Met1-Tyr239). The IgG fraction of the cell culture supernatant was purified by Protein A affinity.
Catalog No. 50-224-8737