Learn More
Aviva Systems Biology Corp PSME3 Antibody - N-terminal region (ARP46142_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46142_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSME3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human PSME3. Alternate names: Ki, PA28-gamma, PA28G, REG-GAMMAApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59246