Learn More
ACROBiosystems ACROBiosystems Human PSME3 / PA28g Protein, His Tag
Catalog No. 501957236
Encompass_Preferred
Recombinant Protein;1MG;Human PSME3, His Tag (PS3-H5142) is expressed from E.coli cells. It contains AA Ala 2 - Tyr 254 (Accession # P61289-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 30.3 kDa. The protein migrates as 28-33 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-195-7236 Supplier ACROBiosystems Supplier No. PS3H51421MG
Recombinant Protein;1MG;Human PSME3, His Tag (PS3-H5142) is expressed from E.coli cells. It contains AA Ala 2 - Tyr 254 (Accession # P61289-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 30.3 kDa. The protein migrates as 28-33 kDa under reducing (R) condition (SDS-PAGE).