Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Psme3 protein, human (N-His) | 1 MG
SDP

Catalog No. 5000290326
Encompass_Preferred

Recombinant human PSME3 protein with an N-terminal His tag, supplied as a 0.22 μm filtered solution in 50 mM Tris, 200 mM NaCl, 40% glycerol (pH 8.0). Expressed in E. coli, this reagent is intended for biochemical and cell biology research into proteasome regulation and protein-protein interactions, including studies of MDM2-p53. Store at -80°C and avoid repeated freeze-thaw cycles.

  • Recombinant human protein with N-terminal His tag.
  • Expressed in E. coli for in vitro research applications.
  • Supplied as filtered glycerol solution in Tris/NaCl, pH 8.0.
  • Molecular weight approximately 28-33 kDa.
  • Stable at -80°C for long-term storage; aliquot recommended to avoid freeze-thaw.
  • Enzyme activity testing in progress; activity not guaranteed.

Catalog No. 50-002-90326 Supplier Medchemexpress LLC Supplier No. HYP76554A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human PSME3 protein with an N-terminal His tag, supplied as a 0.22 μm filtered solution in 50 mM Tris, 200 mM NaCl, 40% glycerol (pH 8.0). Expressed in E. coli, this reagent is intended for biochemical and cell biology research into proteasome regulation and protein-protein interactions, including studies of MDM2-p53. Store at -80°C and avoid repeated freeze-thaw cycles.

  • Recombinant human protein with N-terminal His tag.
  • Expressed in E. coli for in vitro research applications.
  • Supplied as filtered glycerol solution in Tris/NaCl, pH 8.0.
  • Molecular weight approximately 28-33 kDa.
  • Stable at -80°C for long-term storage; aliquot recommended to avoid freeze-thaw.
  • Enzyme activity testing in progress; activity not guaranteed.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.