Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PSME3 protein, human (N-His) | 5 UG
SDP

Catalog No. 5000290087
Encompass_Preferred

Recombinant human PSME3 (proteasome activator complex subunit 3) expressed in E. coli with an N-terminal His tag. Supplied as a lyophilized powder formulated in PBS with trehalose; suitable for biochemical assays and activity measurements.

  • Approximately 33 kDa molecular weight.
  • Lyophilized powder formulated in PBS, pH 7.4, with 8% trehalose.
  • Endotoxin level <1 EU/μg (LAL method).
  • Specific activity ~897.05 pmol/min/μg (suc-LLVY-AMC assay).
  • Reconstitution recommended at ≥100 μg/mL in ddH2O.
  • Store at -20°C; aliquot and freeze at -20°C or -80°C for long-term storage.

Catalog No. 50-002-90087 Supplier Medchemexpress LLC Supplier No. HYP76554A5UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human PSME3 (proteasome activator complex subunit 3) expressed in E. coli with an N-terminal His tag. Supplied as a lyophilized powder formulated in PBS with trehalose; suitable for biochemical assays and activity measurements.

  • Approximately 33 kDa molecular weight.
  • Lyophilized powder formulated in PBS, pH 7.4, with 8% trehalose.
  • Endotoxin level <1 EU/μg (LAL method).
  • Specific activity ~897.05 pmol/min/μg (suc-LLVY-AMC assay).
  • Reconstitution recommended at ≥100 μg/mL in ddH2O.
  • Store at -20°C; aliquot and freeze at -20°C or -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.