Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PSME3 protein, human (N-His) | 96.0% | 10 UG
SDP

Catalog No. 5000292607
Encompass_Preferred

PSME3 Protein, Human (N-His) is a recombinant human proteasome activator subunit 3 expressed in Escherichia coli and supplied lyophilized with an N-terminal His tag. It is intended for research use in biochemical assays, protein interaction studies, and as an assay control; the lyophilized formulation with trehalose supports stability during storage and shipment.

  • N-terminal His tag for affinity purification.
  • Human sequence expressed in E. coli.
  • Approximate molecular weight 33 kDa.
  • Purity >95% as determined by reducing SDS-PAGE.
  • Lyophilized from PBS (pH 7.4) with 8% trehalose.
  • Reconstitute to ≥100 μg/mL; avoid lower concentrations in ddH2O.
  • Store at -20°C; after reconstitution store at 4°C up to 1 week or -20°C for long term.

Catalog No. 50-002-92607 Supplier Medchemexpress LLC Supplier No. HYP76554A10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PSME3 Protein, Human (N-His) is a recombinant human proteasome activator subunit 3 expressed in Escherichia coli and supplied lyophilized with an N-terminal His tag. It is intended for research use in biochemical assays, protein interaction studies, and as an assay control; the lyophilized formulation with trehalose supports stability during storage and shipment.

  • N-terminal His tag for affinity purification.
  • Human sequence expressed in E. coli.
  • Approximate molecular weight 33 kDa.
  • Purity >95% as determined by reducing SDS-PAGE.
  • Lyophilized from PBS (pH 7.4) with 8% trehalose.
  • Reconstitute to ≥100 μg/mL; avoid lower concentrations in ddH2O.
  • Store at -20°C; after reconstitution store at 4°C up to 1 week or -20°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.