Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Psme3 protein, human (N-His) | 96.0% | 100 UG
SDP

Catalog No. 5000292606
Encompass_Preferred

Recombinant human PSME3 (proteasome activator subunit 3) is supplied as an N-terminal His-tagged protein expressed in E. coli and provided as a lyophilized powder intended for research use. The construct corresponds to amino acids M1-Y254 (UniProt P61289) with an approximate molecular weight of 33 kDa.

  • Recombinant human protein corresponding to amino acids M1-Y254.
  • N-terminal 6xHis affinity tag for purification or detection.
  • Expressed in E. coli and lyophilized from PBS (pH 7.4) with 8% trehalose.
  • Purity 96.0% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg as determined by LAL.
  • Reported specific activity 897.05 pmol/min/μg (suc-LLVY-AMC assay).
  • Store at -20°C; after reconstitution stable at 4°C for 1 week or longer at -20°C with carrier.
  • For research use only.

Catalog No. 50-002-92606 Supplier Medchemexpress LLC Supplier No. HYP76554A100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human PSME3 (proteasome activator subunit 3) is supplied as an N-terminal His-tagged protein expressed in E. coli and provided as a lyophilized powder intended for research use. The construct corresponds to amino acids M1-Y254 (UniProt P61289) with an approximate molecular weight of 33 kDa.

  • Recombinant human protein corresponding to amino acids M1-Y254.
  • N-terminal 6xHis affinity tag for purification or detection.
  • Expressed in E. coli and lyophilized from PBS (pH 7.4) with 8% trehalose.
  • Purity 96.0% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg as determined by LAL.
  • Reported specific activity 897.05 pmol/min/μg (suc-LLVY-AMC assay).
  • Store at -20°C; after reconstitution stable at 4°C for 1 week or longer at -20°C with carrier.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.