Learn More
ACROBiosystems Recombinant Protein;Human PSME3 / PA28g Protein ;E.coli;100UG;PS3,PSME3,PA28g,PA28gamma,REG-gamma,PA28-gamma
Supplier:  ACROBiosystems PS3H5142100UG
Encompass_Preferred
Recombinant Protein;Human PSME3 / PA28g Protein ;100UG;Human PSME3, His Tag (PS3-H5142) is expressed from E.coli cells. It contains AA Met 1 - Tyr 254 (Accession # NP_00578).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 30.3 kDa. The protein migrates as 28-33 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8867