Learn More
Aviva Systems Biology Corp PSMG3 Antibody - C-terminal region (ARP60494_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60494_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PSMG3. It was validated on Western Blot. . Alternate names: C7orf48, MGC10911, PAC3ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-287-062