Learn More
Medchemexpress LLC PSN 375963 10mg | 388575-52-8 | 10MG
SDP
Catalog No. 5000242215
Encompass_Preferred
PSN 375963 is a potent GPR119 agonist with EC50s of 8 4 and 7 9 M for human and mouse GPR119 respectively PSN 375963 shows similar potency to the endogenous agonist oleoylethanolamide (OEA)[1 [2
Catalog No. 50-002-42215 Supplier Medchemexpress LLC Supplier No. HY10825810MG
PSN 375963 is a potent GPR119 agonist with EC50s of 8 4 and 7 9 M for human and mouse GPR119 respectively PSN 375963 shows similar potency to the endogenous agonist oleoylethanolamide (OEA)[1 [2