Learn More
ATCC PSN-1; Pancreas Adenocarcinoma; Human (Homo sapiens)
Supplier: ATCC CRMCRL3211
PSN-1 is an epithelial-like cell that was isolated from the pancreas of a 40-year-old, White female with adenocarcinoma. The cell line can be used in testing and calibration in ISO 17025-accredited laboratories, to challenge assay performance, validate or compare test methods, and establish sensitivity, linearity, and specificity during assay validation or implementation.