Learn More
Cayman Chemical Psoala coylfola Bakucol 10mg
SDP
Catalog No. 502952369
Encompass_Preferred
A natural meroterpene that has retinol-like effects on gene expression and properties of the skin; has antioxidant and antibacterial actions; inhibits DNA polymerase and UDP-glucuronosyltransferase 2B7 (IC50 = 41 µM)
Catalog No. 50-295-2369 Supplier Cayman Chemical Supplier No. 1168410 MG
A natural meroterpene that has retinol-like effects on gene expression and properties of the skin; has antioxidant and antibacterial actions; inhibits DNA polymerase and UDP-glucuronosyltransferase 2B7 (IC50 = 41 µM)