Learn More
Selleck Chemical LLC Psoralen S4737-10mg
SDP
Catalog No. 503109745
Encompass_Preferred
Psoralen (Psoralene Ficusin Furocoumarin) is a naturally occurring furocoumarin that intercalates with DNA inhibiting DNA synthesis and cell division
Catalog No. 50-310-9745 Supplier Selleck Chemical LLC Supplier No. S473710MG
Psoralen (Psoralene Ficusin Furocoumarin) is a naturally occurring furocoumarin that intercalates with DNA inhibiting DNA synthesis and cell division