Learn More
Selleck Chemical LLC Psoralen 50mg 66-97-7 Psoralene, Ficusin, Furocoumarin
SDP
Supplier:  Selleck Chemical LLC S473750MG
Encompass_Preferred
Psoralen (Psoralene, Ficusin, Furocoumarin) is a naturally occurring furocoumarin that intercalates with DNA, inhibiting DNA synthesis and cell division. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4261