Learn More
Medchemexpress LLC Psoralen | 66-97-7 | 99.9% | 186.16 | 100 MG
SDP
Catalog No. 5000428939
Encompass_Preferred
Psoralen, also known as Ficusin, is a naturally occurring coumarin compound isolated from the seeds of Fructus Psoraleae. It is intended for research use only.
  • Exhibits anti-cancer activity.
  • Acts as an antioxidant.
  • Demonstrates antidepressant properties.
  • Possesses antibacterial effects.
  • Shows antiviral activity.
Catalog No. 50-004-28939 Supplier Medchemexpress LLC Supplier No. HYN0053100MG
Psoralen, also known as Ficusin, is a naturally occurring coumarin compound isolated from the seeds of Fructus Psoraleae. It is intended for research use only.
  • Exhibits anti-cancer activity.
  • Acts as an antioxidant.
  • Demonstrates antidepressant properties.
  • Possesses antibacterial effects.
  • Shows antiviral activity.