Learn More
Indofine Chemical Psoralen With Hplc, 10 Mg
SDP
Catalog No. 50908852
Encompass_Preferred
PSORALEN with HPLC 10 MG, Coumarin, Ficusin, C11H6O3, MW: 186.16, 98% by HPLC, 66-97-7, MFCD00010520
Catalog No. 50-908-852 Supplier Indofine Chemical Supplier No. 020553S 10 MG
PSORALEN with HPLC 10 MG, Coumarin, Ficusin, C11H6O3, MW: 186.16, 98% by HPLC, 66-97-7, MFCD00010520