Learn More
Medchemexpress LLC Psoralenoside | 905954-17-8 | 5 MG
SDP
Catalog No. 5000417518
Encompass_Preferred
Psoralenoside is a benzofuran glycoside derived from Psoralea corylifolia. It exhibits high binding affinities to histaminergic H1, calmodulin, and voltage-gated L-type calcium channels. This compound also demonstrates estrogen-like activity, accelerates osteoblastic proliferation, and possesses antitumor and antibacterial properties.
  • Exhibits high binding affinities to histaminergic H1, calmodulin, and voltage-gated L-type calcium channels
  • Demonstrates estrogen-like activity
  • Accelerates osteoblastic proliferation
  • Possesses antitumor effects
  • Has antibacterial properties
Catalog No. 50-004-17518 Supplier Medchemexpress LLC Supplier No. HYN75035MG
Psoralenoside is a benzofuran glycoside derived from Psoralea corylifolia. It exhibits high binding affinities to histaminergic H1, calmodulin, and voltage-gated L-type calcium channels. This compound also demonstrates estrogen-like activity, accelerates osteoblastic proliferation, and possesses antitumor and antibacterial properties.
  • Exhibits high binding affinities to histaminergic H1, calmodulin, and voltage-gated L-type calcium channels
  • Demonstrates estrogen-like activity
  • Accelerates osteoblastic proliferation
  • Possesses antitumor effects
  • Has antibacterial properties