Learn More
Cayman Chemical PsoromIc AcId 2.5mg
SDP
Catalog No. 502987041
Encompass_Preferred
An inhibitor of Rab geranylgeranyl transferase (IC50 = 1.3 µM) and P. falciparum FAS-II; cytotoxic to U87MG and KB cancerous cell lines (IC50s = 79.40 and 56.22 mg/L, respectively); active against Plasmodium species (IC50 = 29.2 µM against blood stage P. falciparum)
Catalog No. 50-298-7041 Supplier Cayman Chemical Supplier No. 221682.5 MG
An inhibitor of Rab geranylgeranyl transferase (IC50 = 1.3 µM) and P. falciparum FAS-II; cytotoxic to U87MG and KB cancerous cell lines (IC50s = 79.40 and 56.22 mg/L, respectively); active against Plasmodium species (IC50 = 29.2 µM against blood stage P. falciparum)