Learn More
New England Biolabs, Inc. PspGI – 1000 units
SDP
Catalog No. 50812344
Encompass
A restriction endonuclease that recognizes the sequence ^CCWGG_.
Catalog No. 50-812-344 Supplier New England Biolabs, Inc. Supplier No. R0611S
A restriction endonuclease that recognizes the sequence ^CCWGG_.