Learn More
New England Biolabs, Inc. PspOMI – 1500 units
SDP
Catalog No. 50994919
Encompass
A restriction endonuclease that recognizes the sequence G^GGCC_C.
Catalog No. 50-994-919 Supplier New England Biolabs, Inc. Supplier No. R0653S
A restriction endonuclease that recognizes the sequence G^GGCC_C.