Learn More
New England Biolabs, Inc. PspOMI – 7500 units
SDP
Catalog No. 50995276
Encompass
A restriction endonuclease that recognizes the sequence G^GGCC_C.
Catalog No. 50-995-276 Supplier New England Biolabs, Inc. Supplier No. R0653L
A restriction endonuclease that recognizes the sequence G^GGCC_C.