Learn More
New England Biolabs, Inc. PspXI – 1000 units
SDP
Catalog No. 50995277
Encompass
A restriction endonuclease that recognizes the sequence VC^TCGA_GB.
Catalog No. 50-995-277 Supplier New England Biolabs, Inc. Supplier No. R0656L
A restriction endonuclease that recognizes the sequence VC^TCGA_GB.