Learn More
New England Biolabs, Inc. PspXI – 200 units
SDP
Catalog No. 50994910
Encompass
A restriction endonuclease that recognizes the sequence VC^TCGA_GB.
Catalog No. 50-994-910 Supplier New England Biolabs, Inc. Supplier No. R0656S
A restriction endonuclease that recognizes the sequence VC^TCGA_GB.