Learn More
New England Biolabs, Inc. PstI – 10000 units
SDP
Catalog No. 50812009
Encompass
A restriction endonuclease that recognizes the sequence C_TGCA^G.
Catalog No. 50-812-009 Supplier New England Biolabs, Inc. Supplier No. R0140T
A restriction endonuclease that recognizes the sequence C_TGCA^G.