Learn More
New England Biolabs, Inc. PstI – 50000 units
SDP
Supplier:  New England Biolabs, Inc. R0140M
Encompass
A restriction endonuclease that recognizes the sequence C_TGCA^G.
Catalog No. 50-812-007