Learn More
Andwin Scientific PSTK1000 PETRISTICKERS
SDP
Catalog No. NC0306488
Encompass
The 3 in. diameter petristickers are used to label petri dishes in 32-square grids. They are made with water-resistant ink, substrate and adhesives that can withstand temperatures from 4 to 42 deg.C.
Catalog No. NC0306488 Supplier Andwin Scientific Supplier No. PSTK1000
The 3 in. diameter petristickers are used to label petri dishes in 32-square grids. They are made with water-resistant ink, substrate and adhesives that can withstand temperatures from 4 to 42 deg.C.