Learn More
AVANTOR MASTERFLEX (VWR LLC) PSTNPMP SYSTM 180ML ETFE
Catalog No. 502742113
Encompass
Masterflex Ismatec Reglo Digital Piston Pump Drive with MasterflexLive and Valveless Piston Pump Head, Ceramic/ETFE/Ceramic, 180 mL/min; 115/230 VAC, Avantor
Catalog No. 50-274-2113 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7801865
Masterflex Ismatec Reglo Digital Piston Pump Drive with MasterflexLive and Valveless Piston Pump Head, Ceramic/ETFE/Ceramic, 180 mL/min; 115/230 VAC, Avantor