Learn More
AVANTOR MASTERFLEX (VWR LLC) PSTNPMP SYSTM 45ML ETFE
Catalog No. 502742114
Encompass
Masterflex Ismatec Reglo Digital Piston Pump Drive with MasterflexLive and Valveless Piston Pump Head, Stainless Steel/ETFE/Carbon, 45 mL/min; 115/230 VAC, Avantor
Catalog No. 50-274-2114 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7801862
Masterflex Ismatec Reglo Digital Piston Pump Drive with MasterflexLive and Valveless Piston Pump Head, Stainless Steel/ETFE/Carbon, 45 mL/min; 115/230 VAC, Avantor