Learn More
Medchemexpress LLC Psychosine-d5 | 2260670-12-8 | 99.0% | 466.66 | 1 MG
SDP
Catalog No. 5000348746
Encompass_Preferred
Psychosine-d5 is a deuterium-labeled Psychosine. This compound acts as a substrate for the galactocerebrosidase (GALC) enzyme and is identified as a potential biomarker for Krabbe disease. It is also known as a highly cytotoxic lipid that can induce cell death in various cell types.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-48746 Supplier Medchemexpress LLC Supplier No. HY136490S1MG
Psychosine-d5 is a deuterium-labeled Psychosine. This compound acts as a substrate for the galactocerebrosidase (GALC) enzyme and is identified as a potential biomarker for Krabbe disease. It is also known as a highly cytotoxic lipid that can induce cell death in various cell types.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.