Learn More
Medchemexpress LLC Psychosine-d5 | 2260670-12-8 | 99.0% | 466.66 | 500 UG
SDP
Catalog No. 5000350601
Encompass_Preferred
Psychosine-d5 is a deuterium-labeled form of Psychosine, a substrate of the galactocerebrosidase (GALC) enzyme. It is considered a potential biomarker for Krabbe disease and is a highly cytotoxic lipid known to induce cell death in various cell types. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled compound
  • Potential biomarker for Krabbe disease
  • Highly cytotoxic lipid
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
Catalog No. 50-003-50601 Supplier Medchemexpress LLC Supplier No. HY136490S500UG
Psychosine-d5 is a deuterium-labeled form of Psychosine, a substrate of the galactocerebrosidase (GALC) enzyme. It is considered a potential biomarker for Krabbe disease and is a highly cytotoxic lipid known to induce cell death in various cell types. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled compound
  • Potential biomarker for Krabbe disease
  • Highly cytotoxic lipid
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)