Learn More
Julabo USA Inc Each, Pt100 sensor 2x20mm, 1.5m cable, SS
SDP
Catalog No. 507534120
Encompass_Preferred
Bath Accessories, Julabo, External Pt100 sensor, 20 x 2 mm, stainless steel, 1,5 m cable with 4-pin Lemosa plug. For use with JULABO systems having external Pt100 capability.
Catalog No. 50-753-4120 Supplier Julabo USA Inc Supplier No. 8981006
Bath Accessories, Julabo, External Pt100 sensor, 20 x 2 mm, stainless steel, 1,5 m cable with 4-pin Lemosa plug. For use with JULABO systems having external Pt100 capability.