Learn More
BINDER™ Pt 100 Temperature Sensor
SDP
Catalog No. 08111259 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
BINDER Chambers KT 53
Catalog No. For Use With (Equipment)
08-111-259 BINDER Chambers KT 53
1 options
Catalog No. 08-111-259 Supplier BINDER™ Supplier No. 80120935
Add to Cart
Add to Cart
For Use With: BINDER units KT 53
  • Flexible
  • Additional sensor
  • Interior
Specifications
Description Pt 100 temperature sensor - additional flexible Pt 100, interior, for displaying the temperature on the unit display
For Use With (Equipment) BINDER Chambers KT 53
Material Plastic
Type Temperature Sensor