Description
- Flexible
- Additional sensor
- Interior
Specifications
Specifications
| Description | Pt 100 temperature sensor - additional flexible Pt 100, interior, with external connection incl. LEMO connector (3-pin) |
| For Use With (Equipment) | BINDER Chambers FD 23 |
| Material | Plastic |
| Type | Temperature Sensor |
| Includes | External connection incl. LEMO connector (3-pin) |