Learn More
BINDER™ Pt 100 Temperature Sensor
SDP
Catalog No. 08111815 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
08-111-815 Plastic
1 options
Catalog No. 08-111-815 Supplier BINDER™ Supplier No. 80121176
Add to Cart
Add to Cart
For Use With: FED 400
  • Flexible
  • Additional sensor
  • Interior
Specifications
Description Pt 100 temperature sensor - additional flexible Pt 100, interior, with external connection incl. LEMO connector (3-pin)
For Use With (Equipment) BINDER Chambers FED 400
Material Plastic
Type Temperature Sensor
Includes External connection incl. LEMO connector (3-pin)