Learn More
BINDER™ Pt 100 Temperature Sensor
SDP
Catalog No. 08111816 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
08-111-816 Plastic
1 options
Catalog No. 08-111-816 Supplier BINDER™ Supplier No. 80121618
Add to Cart
Add to Cart
For Use With: BF 56, BF 115, BF 260, BF 720
  • Flexible
  • Additional sensor
  • Interior
Specifications
Description Pt 100 temperature sensor - additional flexible Pt 100, interior, for displaying the temperature on the unit display
For Use With (Equipment) BINDER Chambers BF 56, BF 115, BF 260, BF 720
Material Plastic
Type Temperature Sensor