Learn More
Cayman Chemical PT-141 (acetate), Purity: ≥98%, CAS: 1607799-13-2, 10 mg
SDP
Catalog No. 502910430
Encompass_Preferred
A cyclic peptide agonist of MC4R; increases dopamine release in the mPOA of the hypothalamus in female rats; increases the number of solicitations by ovariectomized estrogen- and progesterone-primed female rats at 100 and 200 µg/kg; induces penile erections in male rats at 50 µg/kg intranasally
Catalog No. 50-291-0430 Supplier Cayman Chemical Supplier No. 3517510 MG
A cyclic peptide agonist of MC4R; increases dopamine release in the mPOA of the hypothalamus in female rats; increases the number of solicitations by ovariectomized estrogen- and progesterone-primed female rats at 100 and 200 µg/kg; induces penile erections in male rats at 50 µg/kg intranasally