Learn More
Medchemexpress LLC HY-12867 10mg , PT-2385 CAS:1672665-49-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1286710MG
Encompass_Preferred
HY-12867 10mg PT-2385 CAS: 1672665-49-4 PT-2385 is a selective HIF-2α inhibitor with a Ki of less than 50 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9044