Learn More
Medchemexpress LLC PT-2385 | 1672665-49-4 | 100.0% | 100 MG
SDP
Catalog No. 5000342939
Encompass_Preferred
PT-2385 is a highly selective HIF-2α inhibitor, demonstrating a Ki of less than 50 nM. It acts as an antagonist specifically targeting HIF-2 over HIF-1, with no activity against HIF-1α. In preclinical studies, PT-2385 has shown significant in vivo activity by inhibiting the expression of HIF-2α regulated genes in a dose-dependent manner. This compound has also been observed to induce rapid, dose-dependent tumor regression in models using 786-O and A498 RCC cell lines, typically when administered at 30 or 100 mg/kg via oral gavage twice daily. Additionally, it contributes to reducing tumor-derived VEGFA protein levels, which in turn diminishes proliferation (Ki67) and angiogenesis (CD-31).
  • Selective HIF-2α inhibitor
  • Inactive against HIF-1α
  • Inhibits HIF-2α regulated gene expression
  • Induces tumor regression in preclinical models
  • Reduces tumor proliferation and angiogenesis
Catalog No. 50-003-42939 Supplier Medchemexpress LLC Supplier No. HY12867100MG
PT-2385 is a highly selective HIF-2α inhibitor, demonstrating a Ki of less than 50 nM. It acts as an antagonist specifically targeting HIF-2 over HIF-1, with no activity against HIF-1α. In preclinical studies, PT-2385 has shown significant in vivo activity by inhibiting the expression of HIF-2α regulated genes in a dose-dependent manner. This compound has also been observed to induce rapid, dose-dependent tumor regression in models using 786-O and A498 RCC cell lines, typically when administered at 30 or 100 mg/kg via oral gavage twice daily. Additionally, it contributes to reducing tumor-derived VEGFA protein levels, which in turn diminishes proliferation (Ki67) and angiogenesis (CD-31).
  • Selective HIF-2α inhibitor
  • Inactive against HIF-1α
  • Inhibits HIF-2α regulated gene expression
  • Induces tumor regression in preclinical models
  • Reduces tumor proliferation and angiogenesis