Learn More
MSC BASF Pressurized Insecticide, Nonrepellent, Broad Spectrum, Commercial Food Areas, 14 oz
Catalog No. 502838353
Encompass
The BASF PT Alpine Pressurized Insecticide is a nonrepellent, broad-spectrum aerosol designed for effective pest control. This convenient, ready-to-use formulation is suitable for use against a wide range of pests, including ants, cockroaches, bed bugs, spiders, and occasional invaders, and is now approved for applications in commercial food handling areas.
  • Broad use label includes commercial food handling areas.
  • Broad spectrum label for use against: ants, cockroaches, bed bugs, spiders, and occasional invaders.
  • Convenient, ready-to-use formulation.
  • A perfect rotational partner in a nonrepellent program.
Catalog No. 50-283-8353 Supplier MSC Supplier No. 74344599
The BASF PT Alpine Pressurized Insecticide is a nonrepellent, broad-spectrum aerosol designed for effective pest control. This convenient, ready-to-use formulation is suitable for use against a wide range of pests, including ants, cockroaches, bed bugs, spiders, and occasional invaders, and is now approved for applications in commercial food handling areas.
  • Broad use label includes commercial food handling areas.
  • Broad spectrum label for use against: ants, cockroaches, bed bugs, spiders, and occasional invaders.
  • Convenient, ready-to-use formulation.
  • A perfect rotational partner in a nonrepellent program.