Learn More
Electron Microscopy Sciences EMS Platinum Aperture with 20 µm hole, 0.165 mm thick
SDP
Catalog No. 50980802
Encompass_Preferred
Platinum apertures are easily cleaned by either an aperture flamer or over a bunsen burner in the open air. We maintain a wide variety of Platinum Drilled Apertures in stock with different configurations suiting all of your needs. Nominal hole diameter: +/- 1 micron. Disc diameter: +/- 10 microns. Disc thickness: +/- 10 microns
Size: 20 µm
O.D.: 4 mm
Thickness: 0.165 mm
Catalog No. 50-980-802 Supplier Electron Microscopy Sciences Supplier No. 45020PT
Platinum apertures are easily cleaned by either an aperture flamer or over a bunsen burner in the open air. We maintain a wide variety of Platinum Drilled Apertures in stock with different configurations suiting all of your needs. Nominal hole diameter: +/- 1 micron. Disc diameter: +/- 10 microns. Disc thickness: +/- 10 microns
Size: 20 µm
O.D.: 4 mm
Thickness: 0.165 mm