Learn More
Electron Microscopy Sciences EMS Platinum Aperture with Spray 500 µm, 12.7 mm O.D.
SDP
Catalog No. 50980820
Encompass_Preferred
Platinum apertures are easily cleaned by either an aperture flamer or over a bunsen burner in the open air. We maintain a wide variety of Platinum Drilled Apertures in stock with different configurations suiting all of your needs. Nominal hole diameter: +/- 1 micron. Disc diameter: +/- 10 microns. Disc thickness: +/- 10 microns
Size: Spray 500 µm
O.D.: 12.7 mm
Catalog No. 50-980-820 Supplier Electron Microscopy Sciences Supplier No. 48500PTS
Platinum apertures are easily cleaned by either an aperture flamer or over a bunsen burner in the open air. We maintain a wide variety of Platinum Drilled Apertures in stock with different configurations suiting all of your needs. Nominal hole diameter: +/- 1 micron. Disc diameter: +/- 10 microns. Disc thickness: +/- 10 microns
Size: Spray 500 µm
O.D.: 12.7 mm