Learn More
Strem, An Ascensus Company CAS# 12080-32-9. 1g. Dichloro(1,5-cyclooctadiene)platinum(II), 99%. MFCD00012413
SDP
Catalog No. 502016737
Encompass
CAS# 12080-32-9. 1g. Dichloro(1,5-cyclooctadiene)platinum(II), 99%. MFCD00012413. Molecular Weight: 374.18. Molecular Formula: PtCl2(1,5-C8H12). Color/form: white to pale yellow solid Strem# 78-0430. http://www.strem.com/catalog/v/78-0430/
Catalog No. 50-201-6737 Supplier Strem, An Ascensus Company Supplier No. 7804301G
CAS# 12080-32-9. 1g. Dichloro(1,5-cyclooctadiene)platinum(II), 99%. MFCD00012413. Molecular Weight: 374.18. Molecular Formula: PtCl2(1,5-C8H12). Color/form: white to pale yellow solid Strem# 78-0430. http://www.strem.com/catalog/v/78-0430/