Learn More
Electron Microscopy Sciences Platinum/Palladium 80, 0.20 mm, Purity 99.95% 50 FT
SDP
Catalog No. 501899993
Encompass_Preferred
High purity, vacuum grade. These metal wires are suitable for scanning and electron microscopy techniques.
Size: 0.20 mm
Purity: 99.95%
Catalog No. 50-189-9993 Supplier Electron Microscopy Sciences Supplier No. 73601
High purity, vacuum grade. These metal wires are suitable for scanning and electron microscopy techniques.
Size: 0.20 mm
Purity: 99.95%