Learn More
METTLER TOLEDO 3P INDUSTRIAL DMi147-SC Plug & Play combined platinum ring electrode
Catalog No. NC1401868
Encompass_Preferred
Combined platinum ring electrode for redox titrations. The electrode has a junction-free bulb glass reference, which cannot be blocked by precipitates. Application examples: Bromatometric, iodometric and cerimetric titrations according to European Pharmacopeia and USP. Determination of Indigo and Hydrosulfite. Peroxide number in oils.
Catalog No. NC1401868 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 51109522
Combined platinum ring electrode for redox titrations. The electrode has a junction-free bulb glass reference, which cannot be blocked by precipitates. Application examples: Bromatometric, iodometric and cerimetric titrations according to European Pharmacopeia and USP. Determination of Indigo and Hydrosulfite. Peroxide number in oils.