Learn More
Adam Equipment Adam Equipment PT Stainless Steel Platform Scale PT312-5S [AE403a] 5000lb / 3000kg capacity x 1lb / 0.5kg readability
Catalog No. NC2612402
Encompass_Preferred
Adam Equipment PT Stainless Steel Platform Scales are ideal for weighing food products, chemicals, pharmaceuticals or mining materials, the PT stainless steel platform features an IP66 rating for protection against water and dust. Dustproof and waterproof load cells are sealed and boast an IP67 rating. An easy-to-clean grade 304 stainless steel pan can tolerate frequent washing, while the sleek brushed surface helps prevent the accumulation of food and dirt particles. The reinforced frame beneath the platform provides strength and support. This PT stainless steel platform is paired with the IP68-rated AE 403 indicator. The AE 403 Indicator features a durable rugged stainless steel design that also protects against water and dust, making it a perfect choice for many industrial applications. Rechargeable battery included for operation almost anywhere.
Catalog No. NC2612402 Supplier Adam Equipment Supplier No. PT 3125S AE403A
Adam Equipment PT Stainless Steel Platform Scales are ideal for weighing food products, chemicals, pharmaceuticals or mining materials, the PT stainless steel platform features an IP66 rating for protection against water and dust. Dustproof and waterproof load cells are sealed and boast an IP67 rating. An easy-to-clean grade 304 stainless steel pan can tolerate frequent washing, while the sleek brushed surface helps prevent the accumulation of food and dirt particles. The reinforced frame beneath the platform provides strength and support. This PT stainless steel platform is paired with the IP68-rated AE 403 indicator. The AE 403 Indicator features a durable rugged stainless steel design that also protects against water and dust, making it a perfect choice for many industrial applications. Rechargeable battery included for operation almost anywhere.