Learn More
Medchemexpress LLC Pt-ttpy | 727721-37-1 | 98.0% | 589.37 g/mol | C22H17Cl2N3Pt | 250 MG
SDP
Catalog No. 5000283995
Encompass_Preferred
Pt-ttpy is a platinum-based metallo-organic complex that selectively binds G-quadruplex DNA structures. It has been reported to induce telomere-related DNA damage, inhibit telomerase activity, and produce chromatin abnormalities during mitosis, making it a useful reagent for studies of telomere biology and anticancer mechanisms.
  • Binds G-quadruplex DNA structures.
  • Induces telomere-related DNA damage in cancer cells.
  • Inhibits telomerase and disrupts telomere function.
  • Causes mitotic chromatin abnormalities such as bridges and double-stranded breaks.
  • Useful as a research reagent for telomere biology and G-quadruplex studies.
  • Provided with high purity suitable for biochemical assays.
Catalog No. 50-002-83995 Supplier Medchemexpress LLC Supplier No. HYW543137250MG
Pt-ttpy is a platinum-based metallo-organic complex that selectively binds G-quadruplex DNA structures. It has been reported to induce telomere-related DNA damage, inhibit telomerase activity, and produce chromatin abnormalities during mitosis, making it a useful reagent for studies of telomere biology and anticancer mechanisms.
  • Binds G-quadruplex DNA structures.
  • Induces telomere-related DNA damage in cancer cells.
  • Inhibits telomerase and disrupts telomere function.
  • Causes mitotic chromatin abnormalities such as bridges and double-stranded breaks.
  • Useful as a research reagent for telomere biology and G-quadruplex studies.
  • Provided with high purity suitable for biochemical assays.