Learn More
Julabo USA Inc Pt100 sensor, 24" x 1/4", PTFE coated, 12' cable with Lemosa connector
SDP
Catalog No. 502068325
Encompass_Preferred
Pt100 temperature sensor, PTFE coated stainless steel for use in corrosive environments; 24" long x 1/4" OD with 12' cable and 4-pin Lemosa connector. For use with any JULABO circulator having external Pt100 sensor capability. Shielded cable.
Catalog No. 50-206-8325 Supplier Julabo USA Inc Supplier No. 8891002
Pt100 temperature sensor, PTFE coated stainless steel for use in corrosive environments; 24" long x 1/4" OD with 12' cable and 4-pin Lemosa connector. For use with any JULABO circulator having external Pt100 sensor capability. Shielded cable.